Dextran, Fluorescein, 70,000 MW, Anionic, Lysine Fixable
Invitrogen™

Dextran, Fluorescein, 70,000 MW, Anionic, Lysine Fixable

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D182225 mg
Catalog number D1822
Price (USD)
404.00
Each
Quantity:
25 mg
Price (USD)
404.00
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

• Label (Ex/Em): Fluorescein (494/521)
• Size: 70,000 MW
• Charge: Anionic
• Fixable: Fixable via Lysine

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeClassic Dyes
Product TypeDextran
Quantity25 mg
Shipping ConditionRoom Temperature
Excitation/Emission494/518 nm
Product LineInvitrogen
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (18)

Citations & References
Abstract
Syntaxin 7 mediates endocytic trafficking to late endosomes.
Authors:Nakamura N, Yamamoto A, Wada Y, Futai M
Journal:J Biol Chem
PubMed ID:10692457
'The lysosome functions are ensured by accurate membrane trafficking in the cell. We found that mouse syntaxin 7 could complement yeast vam3 and pep12 mutants defective in docking/fusion to vacuolar and prevacuolar membranes, respectively. Immunohistochemical studies showed that syntaxin 7 is localized to late endosomes, but not to early endosomes. ... More
A role for phosphoinositide 3-kinase in the completion of macropinocytosis and phagocytosis by macrophages.
Authors:Araki N, Johnson MT, Swanson JA
Journal:J Cell Biol
PubMed ID:8947549
'Phosphoinositide 3-kinase (PI 3-kinase) has been implicated in growth factor signal transduction and vesicular membrane traffic. It is thought to mediate the earliest steps leading from ligation of cell surface receptors to increased cell surface ruffling. We show here that inhibitors of PI 3-kinase inhibit endocytosis in macrophages, not by ... More
Splicing promotes rapid and efficient mRNA export in mammalian cells.
Authors:Valencia P, Dias AP, Reed R,
Journal:Proc Natl Acad Sci U S A
PubMed ID:18287003
'The numerous steps in protein gene expression are extensively coupled to one another through complex networks of physical and functional interactions. Indeed, >25 coupled reactions, often reciprocal, have been documented among such steps as transcription, capping, splicing, and polyadenylation. Coupling is usually not essential for gene expression, but instead enhances ... More
Drosophila Squid/hnRNP helps Dynein switch from a gurken mRNA transport motor to an ultrastructural static anchor in sponge bodies.
Authors:Delanoue R, Herpers B, Soetaert J, Davis I, Rabouille C,
Journal:Dev Cell
PubMed ID:17925228
In Drosophila oocytes, dorso-anterior transport of gurken mRNA requires both the Dynein motor and the heterogeneous nuclear ribonucleoprotein (hnRNP) Squid. We show that gurken transcripts are transported directly on microtubules by Dynein in nonmembranous electron-dense transport particles that also contain Squid and the transport cofactors Egalitarian and Bicaudal-D. At its ... More
Functional protein delivery into neurons using polymeric nanoparticles.
Authors:Hasadsri L, Kreuter J, Hattori H, Iwasaki T, George JM,
Journal:J Biol Chem
PubMed ID:19129199
An efficient route for delivering specific proteins and peptides into neurons could greatly accelerate the development of therapies for various diseases, especially those involving intracellular defects such as Parkinson disease. Here we report the novel use of polybutylcyanoacrylate nanoparticles for delivery of intact, functional proteins into neurons and neuronal cell ... More