Owl™ D2 Wide-Gel Electrophoresis System Combs
Thermo Scientific™
Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTeeth CountThickness (Metric) Teeth
D2-RL-25D25 (2X)1.5 mm
D2-RL-25C25 (2X)1 mm
D2-RL-9D91.5 mm
Catalog number D2-RL-25D
Price (USD)
91.80
Online exclusive
102.00
Save 10.20
Each
Teeth Count:
25 (2X)
Thickness (Metric) Teeth:
1.5 mm
Price (USD)
91.80
Online exclusive
102.00
Save 10.20
Each
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Teeth Count25 (2X)
Thickness (Metric) Teeth1.5 mm
For Use With (Equipment)Owl™ D2 Wide Gel Electrophoresis System
Product LineOwl
TypeRapid Load Comb
Unit SizeEach