Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) - FAQs
Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) - FAQs
View additional product information for Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) - FAQs (D250)
1 product FAQs found
What is the recommended solvent and concentration for Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) (Cat. No. D250) stock solution?
We recommend resuspending Laurdan (6-Dodecanoyl-2-Dimethylaminonaphthalene) (Cat. No. D250) in dimethylformamide (DMF) or acetonitrile at a concentration of 1 mM. The molecular weight for this product is 353.55 g/mol.
The product is stable at room temperature and should be protected from light.