Barnstead™ 1/2 Size B-Pure™ Pressure Gauges
Actual product may vary
Thermo Scientific™
Barnstead™ 1/2 Size B-Pure™ Pressure Gauges
Monitor pressure drop across any of the 10 in. filter cartridges used in the 1/2 size B-Pure™ filter holder with a Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Pressure Gauge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
D2780Gauge Kit, Dual Pressure
D0780Pressure Gauge, Single
Catalog number D2780
Price (USD)
422.10
Online Exclusive
469.00
Save 46.90 (10%)
Each
Description:
Gauge Kit, Dual Pressure
Price (USD)
422.10
Online Exclusive
469.00
Save 46.90 (10%)
Each
Monitor pressure drop across any of the 10 in. filter cartridges used in the B-Pure™ 1/2 size filter holder with a Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Pressure Gauge.
  • Attach to any B-Pure system 0 to 160psi pressure gauge, allowing for monitoring inlet and outlet water pressure.
  • Monitor pressure drop across any of the 10 in. filter cartridges used in the 1/2 size B-Pure filter holder.
  • Pressure drop of greater than 10 psi indicates need for filter change.
Specifications
DescriptionGauge Kit, Dual Pressure
For Use With (Equipment)Any B-Pure System
TypePressure Gauge, Dual
Unit SizeEach