DiSC<sub>3</sub>(5) (3,3'-Dipropylthiadicarbocyanine Iodide)
Invitrogen™
DiSC3(5) (3,3'-Dipropylthiadicarbocyanine Iodide)
La sonda potenciométrica, DiSC3(5) es una carbocianina con una cola de alquilo corta (C3). Este tinte catiónico se acumula enMás información
Have Questions?
Número de catálogoCantidad
D306100 mg
Número de catálogo D306
Precio (MXN)
-
Cantidad:
100 mg
La sonda potenciométrica, DiSC3(5) es una carbocianina con una cola de alquilo corta (C3). Este tinte catiónico se acumula en las membranas hiperpolarizadas y se transcribe en la bicapa lipídica.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Método de detecciónFluorescente
Cantidad100 mg
Condiciones de envíoTemperatura ambiente
Localización subcelularMembranas celulares y lípidos
ColorRojo
Para utilizar con (equipo)Microscopio de fluorescencia
Tipo de productoDiSC3(5)
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente y proteger de la luz.
Preguntas frecuentes
I stained my cells with a lipophilic cyanine dye, like DiI, but the signal was lost when I tried to follow up with antibody labeling. Why?
Since these dyes insert into lipid membranes, any disruption of the membranes leads to loss of the dye. This includes permeabilization with detergents like Triton X-100 or organic solvents like methanol. Permeabilization is necessary for intracellular antibody labeling, leading to loss of the dye. Instead, a reactive dye such as CFDA SE should be used to allow for covalent attachment to cellular components, thus providing for better retention upon fixation and permeabilization.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How long does it take for lipophlic tracers to transport along the membrane? How much faster are the FAST lipophilic dyes?
The transport is fairly slow, around 6 mm/day in live tissue and slower in fixed tissue, so diffusion of lipophilic carbocyanine tracers from the point of their application to the terminus of a neuron can take several days to weeks The FAST DiO and DiI analogs (which have unsaturated alkyl tails) can improve transport rate by around 50%.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Which form of the lipophilic tracers (DiO, DiI, DiD, etc) should I use?
Select the dye that is compatible with your available excitation source(s) and emission filter set/channels. The solid, paste and crystal forms can be applied directly to neurons in tissues. For labeling cells in culture or microinjection, the lipophilic dyes in solution or solid form can be used.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I want to label two cell populations and then perform a cell fusion assay. Which reagents are best for imaging this?
Lipophilic cyanine dyes are preferred for this sort of assay, since they insert into cellular membranes and then, upon fusion, are shared by the fused cells as the membranes are shared. For example, one cell population can be labeled with DiI (orange-red) and another cell population can be labeled with DiO (green), and when the cells fuse, the combined color appears yellow (when imaged with a dual-bandpass filter set).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (222)
Citations & References
Abstract
Quantitation of corneal endothelial potentials using a carbocyanine dye.
Authors:Graves C, Sachs G
Journal:Biochim Biophys Acta
PubMed ID:6977376
The carbocyanine dye, diS-C3-(5) was used to quantitate the plasma membrane potential of the bullfrog corneal endothelium. It was shown that valinomycin hyperpolarized the endothelial cell and that in the presence of the ionophore the membrane potential largely reflected the K+ equilibrium potential. Using calibration curves constructed by changing medium ... More
Visualization of the intracellular behavior of HIV in living cells.
Authors:McDonald D, Vodicka MA, Lucero G, Svitkina TM, Borisy GG, Emerman M, Hope TJ
Journal:J Cell Biol
PubMed ID:12417576
'To track the behavior of human immunodeficiency virus (HIV)-1 in the cytoplasm of infected cells, we have tagged virions by incorporation of HIV Vpr fused to the GFP. Observation of the GFP-labeled particles in living cells revealed that they moved in curvilinear paths in the cytoplasm and accumulated in the ... More
Measurement of membrane potential in Bacillus subtilis: a comparison of lipophilic cations, rubidium ion, and a cyanine dye as probes.
Authors:Zaritsky A, Kihara M, Macnab RM
Journal:J Membr Biol
PubMed ID:6796695
Monitoring the effect of an anti-cancer drug on RPMI 4788 cells by a membrane potential probe, dis-C3-(5).
Authors:Iwagaki H, Fuchimoto S, Shiiki S, Miyake M, Orita K
Journal:J Med
PubMed ID:2504864
Use of intracellular calcium and membrane potential fluorescent indicators in neuroendocrine cells.
Authors:Capponi AM, Lew PD, Schlegel W, Pozzan T
Journal:Methods Enzymol
PubMed ID:3713520