Dextran, Texas Red™, 3000 MW, Lysine Fixable
Invitrogen™

Dextran, Texas Red™, 3000 MW, Lysine Fixable

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D332810 mg
Catalog number D3328
Price (USD)
427.65
Online exclusive
450.00
Save 22.35
Each
Quantity:
10 mg
Price (USD)
427.65
Online exclusive
450.00
Save 22.35
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

• Label (Ex/Em): Texas Red™ (595/615)
• Size: 3,000 MW
• Charge: Zwitterionic
• Fixable: Fixable via Lysine

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeClassic Dyes
Product TypeDextran
Quantity10 mg
Shipping ConditionRoom Temperature
Excitation/Emission595/615 nm
Product LineTexas Red
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I can't see the structural details of neurons when I inject my fluorescent dextran. What can I do to improve the detailed structure?

If you want to see the most detailed structure you should use the low molecular weight conjugated dextrans such as the 3,000 MW dextrans.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Why isn't my fluorescently conjugated dextran signal retained after fixation?

Ensure that the dextran you are using is the fixable form (i.e., contains a primary amine). Dextrans that do not contain a primary amine will not be fixed. Another factor could be that the concentration of the dextran is too low, and the concentration use can be increased up to 10 mg/mL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the charges of the dextrans?

We do not determine the net charge of the dextran conjugates. The net charge depends on the fluorophore used to label the dextran and the method of preparing the conjugate. We label some dextrans as neutral or anionic based on the fluorophore used, however the net charge of the dextran may not always be the same as the dye. The Alexa Fluor, Cascade Blue, Lucifer Yellow, fluorescein, and Oregon Green dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic Rhodamine B, tetramethylrhodamine and Texas Red dyes are essentially neutral.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What size dextran is best for neuronal tracing?

Dextrans with molecular weights from 3,000 to 70,000 have been used, however the 3,000 and 10,000 MW dextrans are most commonly used for neuronal tracing. The 3,000 MW dextrans are used for more detailed tracing of fine neuronal projections, investigating gap junctions, and diffuse more quickly; while the 10,000 MW dextrans have slower distribution, longer cellular retention, and do not cross gap junctions.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you have a neuronal tracing protocol?

The NeuroTrace BDA-10,000 Neuronal Tracer Kit (Cat. No. N7167) manual has a good protocol for injection procedures and neuronal tracing using the10,000 MW lysine-fixable biotin dextran amine (BDA). This protocol could potentially be applied to other fluorescent dextrans.

Please review Tables 1a and 1b on pages 4 and 5 - https://tools.thermofisher.com/content/sfs/manuals/mp07167.pdf

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (28)

Citations & References
Abstract
Improved retrograde axonal transport and subsequent visualization of tetramethylrhodamine (TMR) -dextran amine by means of an acidic injection vehicle and antibodies against TMR.
Authors:Kaneko T, Saeki K, Lee T, Mizuno N
Journal:J Neurosci Methods
PubMed ID:8740593
'We studied the ability of various dextran amines (DA) to retrogradely label cortical neurons to the full extent of their dendritic configurations. Corticothalamic neurons were labeled by pressure injection of DA into the ventrobasal thalamic nuclei of the rat brain. Of fluorescein-, Texas Red-, Cascade Blue- and tetramethylrhodamine (TMR)-DAs of ... More
Intracellular fluid flow in rapidly moving cells.
Authors:Keren K, Yam PT, Kinkhabwala A, Mogilner A, Theriot JA,
Journal:Nat Cell Biol
PubMed ID:19767741
'Cytosolic fluid dynamics have been implicated in cell motility because of the hydrodynamic forces they induce and because of their influence on transport of components of the actin machinery to the leading edge. To investigate the existence and the direction of fluid flow in rapidly moving cells, we introduced inert ... More
Kinesin dependent, rapid, bi-directional transport of ER sub-compartment in dendrites of hippocampal neurons.
Authors:Bannai H, Inoue T, Nakayama T, Hattori M, Mikoshiba K
Journal:J Cell Sci
PubMed ID:14676272
'Although spatially restricted Ca2+ release from the endoplasmic reticulum (ER) through intracellular Ca2+ channels plays important roles in various neuronal activities, the accurate distribution and dynamics of ER in the dendrite of living neurons still remain unknown. To elucidate these, we expressed fluorescent protein-tagged ER proteins in cultured mouse hippocampal ... More
In vivo diffusion analysis with quantum dots and dextrans predicts the width of brain extracellular space.
Authors:Thorne RG, Nicholson C
Journal:Proc Natl Acad Sci U S A
PubMed ID:16567637
'Diffusion within the extracellular space (ECS) of the brain is necessary for chemical signaling and for neurons and glia to access nutrients and therapeutics; however, the width of the ECS in living tissue remains unknown. We used integrative optical imaging to show that dextrans and water-soluble quantum dots with Stokes-Einstein ... More
Intravital multiphoton microscopy of dynamic renal processes.
Authors:Molitoris BA, Sandoval RM
Journal:Am J Physiol Renal Physiol
PubMed ID:15883167
'Recent advances in microscopy and optics, computer sciences, and the available fluorophores used to label molecules of interest have empowered investigators to utilize intravital two-photon microscopy to study the dynamic events within the functioning kidney. This emerging technique enables investigators to follow functional and structural alterations with subcellular resolution within ... More