BODIPY&trade; FL C<sub>5</sub>-Ceramide (<i>N</i>-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene-3-Pentanoyl)Sphingosine)
Invitrogen™
BODIPY™ FL C5-Ceramide (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosine)
BODIPY™ FL C5 ceramide can be used in sphingolipid transport and metabolism studies.Read more
Have Questions?
Catalog NumberQuantity
D3521250 μg
Catalog number D3521
Price (USD)
449.00
Each
Quantity:
250 μg
Price (USD)
449.00
Each
BODIPY™ FL C5 ceramide can be used in sphingolipid transport and metabolism studies.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialSphingolipids
Recommended StorageStore in freezer (-5 to -30°C) and protect from light.
Physical FormSolid
Product LineBODIPY
Quantity250 μg
Unit SizeEach
Citations & References (128)
Citations & References
Abstract
Time-lapse analysis and mathematical characterization elucidate novel mechanisms underlying muscle morphogenesis.
Authors:Snow CJ, Goody M, Kelly MW, Oster EC, Jones R, Khalil A, Henry CA,
Journal:PLoS Genet
PubMed ID:18833302
'Skeletal muscle morphogenesis transforms short muscle precursor cells into long, multinucleate myotubes that anchor to tendons via the myotendinous junction (MTJ). In vertebrates, a great deal is known about muscle specification as well as how somitic cells, as a cohort, generate the early myotome. However, the cellular mechanisms that generate ... More
Two-photon fluorescence absorption and emission spectra of dyes relevant for cell imaging.
Authors:Bestvater F, Spiess E, Stobrawa G, Hacker M, Feurer T, Porwol T, Berchner-Pfannschmidt U, Wotzlaw C, Acker H
Journal:J Microsc
PubMed ID:12423261
'Two-photon absorption and emission spectra for fluorophores relevant in cell imaging were measured using a 45 fs Ti:sapphire laser, a continuously tuneable optical parametric amplifier for the excitation range 580-1150 nm and an optical multichannel analyser. The measurements included DNA stains, fluorescent dyes coupled to antibodies as well as organelle ... More
Use of fluorescent dyes for measurement and localization of organelles associated with Ca2+ store release in human neutrophils.
Authors:Davies EV, Blanchfield H, Hallett MB
Journal:Cell Biol Int
PubMed ID:9693835
'Fura-2 and its lipid analogue, FFP-18, were used to measure changes in cytosolic free Ca2+ concentration within human neutrophils. Whereas fura-2 was employed to monitor cytosolic Ca2+ increases throughout the cytosol, FFP-18 was used to monitor Ca2+ changes only near the membrane. This latter probe was incorporated into the plasma ... More
Changes in architecture of the Golgi complex and other subcellular organelles during myogenesis.
Authors:Ralston E
Journal:J Cell Biol
PubMed ID:7678420
'Myogenesis involves changes in both gene expression and cellular architecture. Little is known of the organization, in muscle in vivo, of the subcellular organelles involved in protein synthesis despite the potential importance of targeted protein synthesis for formation and maintenance of functional domains such as the neuromuscular junction. A panel ... More
Porphyrin-retinamides: synthesis and cellular studies.
Authors:Sibrian-Vazquez M, Jensen TJ, Vicente MG
Journal:Bioconjug Chem
PubMed ID:17518439
'A series of four porphyrin-retinamides containing either all-trans- or 13-cis-retinoid acid residues, directly linked to the para-phenyl position of meso-tetraphenylporphyrin or via a low-molecular-weight PEG spacer, have been synthesized. The biological properties of these conjugates were evaluated in a model cell line, human HEp2, and in neuroblastoma SK-N-DZ cells, which ... More