BODIPY&trade; 558/568 C<sub>12</sub> (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene-3-Dodecanoic Acid)
Invitrogen™

BODIPY™ 558/568 C12 (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)

The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
Have Questions?
Catalog NumberQuantity
D38351 mg
Catalog number D3835
Price (USD)
150.88
Special offer
Online exclusive
Ends: 31-Dec-2026
184.00
Save 33.12
Each
Quantity:
1 mg
Price (USD)
150.88
Special offer
Online exclusive
Ends: 31-Dec-2026
184.00
Save 33.12
Each

The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration up to 1 mM.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialFatty Acids
Excitation/Emission559/568 nm
Recommended StorageStore in freezer (-5°C to -30°C) and protect from light.
Physical FormSolid
Product LineBODIPY
Quantity1 mg
Unit SizeEach

Citations & References (19)

Citations & References
Abstract
Manipulating FRET with polymeric vesicles: development of a "mix-and-detect" type fluorescence sensor for bacterial toxin.
Authors:Ma G, Cheng Q
Journal:Langmuir
PubMed ID:16863214
'A simple "mix-and-detect" type of fluorescence sensor for cholera toxin (CT) is reported. The sensor consists of a BODIPY lipid dye and polydiacetylene (PDA) vesicles and utilizes the lipid insertion and FRET mechanism to offer a direct and fluorescence "turn-on" detection of the analyte. BODIPY conjugated GM1, dissolved in a ... More
Photolysis of caged phosphatidic acid induces flagellar excision in Chlamydomonas.
Authors:Goedhart J, Gadella TW
Journal:Biochemistry
PubMed ID:15065870
'Phosphatidic (PtdOH) acid formation is recognized as an important step in numerous signaling pathways in both plants and mammals. To study the role of this lipid in signaling pathways, it is of major interest to be able to increase the amount of this lipid directly. Therefore, "caged" PtdOH was synthesized, ... More
The emergence of competition between model protocells.
Authors:Chen IA, Roberts RW, Szostak JW
Journal:Science
PubMed ID:15353806
The transition from independent molecular entities to cellular structures with integrated behaviors was a crucial aspect of the origin of life. We show that simple physical principles can mediate a coordinated interaction between genome and compartment boundary, independent of any genomic functions beyond self-replication. RNA, encapsulated in fatty acid vesicles, ... More
Altered lipid droplet dynamics in hepatocytes lacking triacylglycerol hydrolase expression.
Authors:Wang H, Wei E, Quiroga AD, Sun X, Touret N, Lehner R,
Journal:Mol Biol Cell
PubMed ID:20410140
Lipid droplets (LDs) form from the endoplasmic reticulum (ER) and grow in size by obtaining triacylglycerols (TG). Triacylglycerol hydrolase (TGH), a lipase residing in the ER, is involved in the mobilization of TG stored in LDs for the secretion of very-low-density lipoproteins. In this study, we investigated TGH-mediated changes in ... More
Fluorogenic substrates for high-throughput measurements of endothelial lipase activity.
Authors:Mitnaul LJ, Tian J, Burton C, Lam MH, Zhu Y, Olson SH, Schneeweis JE, Zuck P, Pandit S, Anderson M, Maletic MM, Waddell ST, Wright SD, Sparrow CP, Lund EG
Journal:J Lipid Res
PubMed ID:17090660
Endothelial lipase (EL) has been shown to be a critical determinant for high density lipoprotein cholesterol levels in vivo; therefore, assays that measure EL activity have become important for the discovery of small molecule inhibitors that specifically target EL. Here, we describe fluorescent Bodipy-labeled substrates that can be used in ... More