Barnstead™ Mega-Pure™ Accessories
Actual product may vary
Thermo Scientific™
Barnstead™ Mega-Pure™ Accessories
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure™ Glass Still, including storage bottles and deionization cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
D400499High Capacity Cartridge
440236Dual Solenoid, MP-1A (240V), MP-3, MP-6A, and MP-11A
D400377High Purity Cartridge
RY798X2ADual Solenoid, MP-1A (120V)
Catalog number D400499
Price (USD)
465.00
Each
Description:
High Capacity Cartridge
Price (USD)
465.00
Each
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure Glass Still. Accessories include dual solenoid, deionization cartridges, storage solutions, single cartridge deionizer, dual cartridge deionizer, deionizer accessories and cartridges, storage bottles and automatic collection system.
Specifications
DescriptionHigh Capacity Cartridge
For Use With (Equipment)MP-12, D1 or D2 Deionizers
TypeCartridge, High Capacity
Unit SizeEach