Barnstead™ Nanopure™ Accessories and Consumables
Actual product may vary
Thermo Scientific™
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D50281Deionized Water Feed (Ultralow Organics, Type I Water)
D50280R/O and Distilled Feed (Ultralow Organics, Type I Water)
D50282R/O and Distilled Feed (Low Organics, Type I Water)
D50283Deionized Water Feed (Low Organics, Type I Water)
D8964Low Water Level Pump Protector
LMX26UV Lamp (TOC)
Catalog number D50281
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each
Type:
Deionized Water Feed (Ultralow Organics, Type I Water)
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
For Use With (Equipment)Nanopure Water Purifications System
TypeDeionized Water Feed (Ultralow Organics, Type I Water)
Unit SizeEach