Barnstead™ Nanopure™ Accessories and Consumables
Thermo Scientific™
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D50283Deionized Water Feed (Low Organics, Type I Water)
D50280R/O and Distilled Feed (Ultralow Organics, Type I Water)
D50281Deionized Water Feed (Ultralow Organics, Type I Water)
D50282R/O and Distilled Feed (Low Organics, Type I Water)
D8964Low Water Level Pump Protector
LMX26UV Lamp (TOC)
Catalog number D50283
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each
Type:
Deionized Water Feed (Low Organics, Type I Water)
Price (USD)
1,377.00
Online exclusive
1,530.00
Save 153.00
Each
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
For Use With (Equipment)Nanopure Water Purifications System
TypeDeionized Water Feed (Low Organics, Type I Water)
Unit SizeEach