Barnstead™ Easypure™ RoDi Accessories and Consumables
Actual product may vary
Thermo Scientific™
Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
D7034Empty Cartridges
AY1332X1External Booster Pump
AY1332X2Pre-filtration Accessory Kit
CV742X5AVentgard Reservoir Cap
D13324Wall Mounting Bracket
D502132Start-up Kit
D502133Cartridge Kit
FL1332X1Reverse Osmosis Membrane
Catalog number D7034
Price (MXN)
-
Description:
Empty Cartridges
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
DescriptionEmpty Cartridges
For Use With (Equipment)Barnstead Easypure RoDi
TypeCartridge, Empty
Unit SizeEach