Dextran, Oregon Green™ 488; 70,000 MW, Anionic
Invitrogen™

Dextran, Oregon Green™ 488; 70,000 MW, Anionic

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D7172
also known as D-7172
5 mg
Catalog number D7172
also known as D-7172
Price (USD)
379.00
Each
Quantity:
5 mg
Price (USD)
379.00
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

• Label (Ex/Em): Oregon Green™ 488 (496/524)
• Size: 70,000 MW
• Charge: Anionic
• Fixable: Nonfixable

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeOther Label(s) or Dye(s)
Product TypeDextran
Quantity5 mg
Shipping ConditionRoom Temperature
Excitation/Emission496/524 nm
Product LineOregon Green
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (4)

Citations & References
Abstract
Fluorescent calcium indicators: subcellular behavior and use in confocal imaging.
Authors:O'Malley DM, Burbach BJ, Adams PR
Journal:Methods Mol Biol
PubMed ID:10231797
Activation of lysosomal function during dendritic cell maturation.
Authors:Trombetta ES, Ebersold M, Garrett W, Pypaert M, Mellman I
Journal:Science
PubMed ID:12610307
In response to a variety of stimuli, dendritic cells (DCs) transform from immature cells specialized for antigen capture into mature cells specialized for T cell stimulation. During maturation, the DCs acquire an enhanced capacity to form and accumulate peptide-MHC (major histocompatibility complex) class II complexes. Here we show that a ... More
Role of CD4+ T Helper Cells in the Development of BAC-Induced Dry Eye Syndrome in Mice.
Authors:
Journal:Invest Ophthalmol Vis Sci
PubMed ID:33492357
Structural and Functional Characterization of Fibronectin in Extracellular Vesicles From Hepatocytes.
Authors:
Journal:Front Cell Dev Biol
PubMed ID:33816490