Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Actual product may vary
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Thermo Scientific™
Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Accurately dispense media and achieve powerful, quiet, reliable homogenization with maximum productivity in your sample preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypePackaging Quantity
DB5003ABag Clips50 clips
DB4001ADispensing Platform1 Ea.
DB4002ATrocar1 Ea.
DB4003ABroth Bag Stand1 Ea.
DB4004ATube Holding Arm1 Ea.
DB4005ABottle1 Ea.
DB4006ACalibration Weight1 Ea.
DB4007AFoot Pedal1 Ea.
DB4008APrinter1 Ea.
DB4009ANozzles5 pieces/Bag
DB4010AWeight1 Ea.
DB4016APrinter Stickers2 Rolls, 980/Roll
DB4017APrinter Roll2 Rolls
DB4018ADispensing Assembly1 Ea.
DB4019ATubing Set1 Ea.
DB4020APump1 unit
DB5001ABag Stand1 Ea.
DB5002ABag Stand1 Rack
Catalog number DB5003A
Price (USD)
-
Product Type:
Bag Clips
Packaging Quantity:
50 clips
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
The easy-to-use Diluflux Gravimetric Dilutor allows the simultaneous use of up to two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted on the unit. The reliable and powerful Homogenizer Laboratory Blender enables rapid and efficient sample homogenization with a small footprint and quiet operation. The accessories listed here enable:
  • Alternative receptacles (bottles instead of bags)
  • Alternative media sources (broth bags/bottles)
  • Additional/replacement bag stands for ease of preparation
  • Bag storage for incubation
  • Sample labeling
  • Maintenance with replacement parts
Contact us to arrange a demonstration of the Dilutor, the Blender, and/or its accessories.
Not all products are available for sale in all territories. Please inquire.
For research use only.
Specifications
DescriptionBag clips for 400 mL bags
For Use With (Equipment)Diluflux Diluter, Homogenizer, Bag Clips
Packaging Quantity50 clips
Product TypeBag Clips
Unit SizeEach