Product Image
Thermo Scientific™
Oxoid™ ONPG Disc
Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.
Have Questions?
Catalog NumberQuantity
DD0013T50 Discs
Catalog number DD0013T
Price (USD)
-
Quantity:
50 Discs
Thermo Scientific™ Oxoid ONPG Disc is used for rapid detection of beta-galactosidase activity.
Specifications
FormatDisk
IncludesONPG Disk
Quantity50 Discs
TypeONPG Disk
Unit SizeEach
Contents & Storage
2°C to 8°C