FitBag™ Half Fraser Broth
FitBag™ Half Fraser Broth
FitBag™ Half Fraser Broth
FitBag™ Half Fraser Broth
Thermo Scientific™

FitBag™ Half Fraser Broth

Shorten your workflow with easy to prepare 2.7L, 4.5L or 9L media bags: ready in 5 to 10 minutes, while segmented structure allows pre-filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
DF0895B20 x 4.5 L
DF0895A30 x 2.7 L
DF0895C10 x 9.0 L
Catalog number DF0895B
Price (EUR)
-
Quantity:
20 x 4.5 L

Save time in preparing selective enrichment for the isolation of Listeria spp. from food and environmental samples using Thermo Scientific™ FitBag™ Half Fraser Broth. The irradiated medium is supplied in lightweight, transparent, plastic bags—just add water and the medium is ready for use. The bag's segmented structure allows pre-filling so that you can reconstitute in two minutes when needed. Available in 2.7L, 4.5L or 9L sizes to suit your workflow, medium can be dispensed through the outlet tube as required.

Shorten your workflow with FitBag UVM1 Broth. FitBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

For Laboratory Use
For Laboratory Use
1. Fraser J.A. and Sperber W.H. (1988) J. Food Protect. 51, No.10, 762-765.
Specifications
DescriptionFitBag Half Fraser Broth
FormatConvenience Format
Quantity20 x 4.5 L
Shelf Life2 years (rehydrated 10 Days)
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
10°C to 30°C