FitBag™ 24 Listeria Enrichment Broth (LEB) Complete
Actual product may vary
FitBag™ 24 Listeria Enrichment Broth (LEB) Complete
FitBag™ 24 Listeria Enrichment Broth (LEB) Complete
FitBag™ 24 Listeria Enrichment Broth (LEB) Complete
Thermo Scientific™

FitBag™ 24 Listeria Enrichment Broth (LEB) Complete

Shorten your workflow with easy to prepare 2.7L, 4.5L or 9L media bags: ready in five to 10 minutes, while segmented structure allows pre-filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
DF1107B20 x 4.5 L
DF1107A30 x 2.7 L
DF1107C10 x 9 L
Catalog number DF1107B
Price (USD)
-
Quantity:
20 x 4.5 L

Save time in preparing selective enrichment medium for Listeria monocytogenes and other Listeria spp. from food and environmental samples using Thermo Scientific™ FitBag™ 24 Listeria Enrichment Broth (LEB) Complete. The irradiated medium is supplied in lightweight, transparent, plastic bags—just add water and the medium is ready for use. The bag's segmented structure allows pre-filling so that you can reconstitute in two minutes when needed. Available in 2.7L, 4.5L or 9L sizes to suit your workflow, medium can be dispensed through the outlet tube as required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which FitBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

Reduces waste: unprepared FitBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, FitBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

For Laboratory Use
For Laboratory Use
Specifications
DescriptionFitBag 24 Listeria Enrichment Broth (LEB)
FormatConvenience Format
Media TypeListeria Enrichment Broth
Quantity20 x 4.5 L
Shelf Life2 years (rehydrated 10 Days)
Product TypeNutrient Broth
Unit SizeEach
Contents & Storage
10°C to 30°C