MAS™ DOA TOTAL Controls
MAS™ DOA TOTAL Controls
Actual product may vary
Thermo Scientific™
MAS™ DOA TOTAL Controls
Monitor immunoassay screening procedures with multi-constituent urine toxicology controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
DOAT-11
DOAT-22
DOAT-33
DOAT-44
DOAT-55
DOAT-66
DOAT-MPMultipack-6 levels
Catalog number DOAT-1
Price (USD)
-
Request A Quote
Level:
1
Monitor immunoassay screening procedures with liquid, ready-to-use Thermo Scientific™ MAS™ DOA TOTAL Controls. These are multi-constituent urine toxicology controls, offering 19 analytes with four distinct levels at drug concentrations 25% below and above commonly used screening and SAMHSA cutoffs. A drug-free level and high positive level are also available (six levels total).
  • Human urine based matrix
  • GC/MS confirmation values available
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionDOA TOTAL Control
Level1
Packaging6 Pk.
Quantity18 mL
Shelf Life24 Months
TypeControl
Detectable AnalytesBenzoylecgonine, Buprenorphine, Cotinine, d-Methamphetamine, EDDP, Ethanol, Ethyl Glucuronide (EtG), LSD, Methadone, Methaqualone, Morphine-free, Nitrazepam, Nortriptyline, Oxazepam, Oxycodone, Phencyclidine (PCP), Propoxyphene (PPX), Secobarbital, 11-nor-9-COOH-D9-THC
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C