| DR0703G | Full specifications | Each | PathoDxtra Strep Group C Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group C Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0704G | Full specifications | Each | PathoDxtra Strep Group D Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group D Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0705G | Full specifications | Each | PathoDxtra Strep Group F Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group F Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0701G | Full specifications | Each | PathoDxtra Strep Group A Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group A Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0710M | Full specifications | Each | PathoDxtra Strep Grouping Reagent Set | 1 Set | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Grouping Reagent Set | Detectable Analytes | beta-haemolytic streptococci | Type | Reagents for Streptococcus Testing | For Use With (Application) | Enzyme Immunoassay Detection | Includes | Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 1 Set | Unit Size | Each |
|
| DR0707G | Full specifications | Each | PathoDxtra Polyvalent Positive Control | 2.8 mL | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Polyvalent Positive Control | Detectable Analytes | beta-haemolytic streptococci | Type | Positive Control | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 2.8 mL | Unit Size | Each |
|
| DR0702G | Full specifications | Each | PathoDxtra Strep Group B Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group B Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0706G | Full specifications | Each | PathoDxtra Strep Group G Latex | 60 Tests | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Strep Group G Latex | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests | Unit Size | Each |
|
| DR0709M | Full specifications | Each | PathoDxtra Extraction Reagents | 1 Set | Request A Quote |
Control Sets | Positive | Description | PathoDxtra Extraction Reagents | Detectable Analytes | beta-haemolytic streptococci | Type | Reagents for Streptococcus Testing | For Use With (Application) | Enzyme Immunoassay Detection | Includes | Extraction Reagent 1, 4mL; Extraction Reagent 2, 4mL; Extraction Reagent 3, 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 1 Set | Unit Size | Each |
|
| DR0700M | Full specifications | Each | PathoDxtra™ Strep Grouping Kit | 60 Tests/Kit | Request A Quote |
Control Sets | Positive | Description | PathoDxtra™ Strep Grouping Kit | Detectable Analytes | beta-haemolytic streptococci | Type | Streptococcus Test Kit | For Use With (Application) | Enzyme Immunoassay Detection | Includes | 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1, 4mL; Extraction Reagent 2, 4mL; Extraction Reagent 3, 2 x 10mL; Mixing Sticks; Disposable Reaction Cards | Material | Polyvalent | Organism Group | Group A, B, C, D, F and G | Quantity | 60 Tests/Kit | Unit Size | Each |
|