Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Nalgene™ Reusable Filter Funnel with Clamp
Thermo Scientific™
Nalgene™ Reusable Filter Funnel with Clamp
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Have Questions?
Catalog NumberCapacity (Metric)
DS0315-0047250 mL
Catalog number DS0315-0047
Price (USD)
254.15
Online exclusive
314.00
Save 59.85
Each
Capacity (Metric):
250 mL
Price (USD)
254.15
Online exclusive
314.00
Save 59.85
Each
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
WARNING: Cancer – www.P65Warnings.ca.gov
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Specifications
AutoclavableAutoclavable
Diameter (Metric)Top47 mm
For Use With (Application)High-throughput particle and microbiological QC testing
IncludesRugged Polysulfone Funnel, Polypropylene Copolymer Cover with 3 ports, Friction-fit TPE Caps (3 caps), Support Plate for 47 mm membrane, Silicone Gasket, No. 8 Stopper with 0.5625 in. hole, Aluminum Clamp
Quantity1 Ea.
Capacity (Metric)250 mL
MaterialPSU
Product TypeFiltering Funnel
Unit SizeEach