Product Image
Thermo Scientific™
Nalgene™ Rubber Stopper, for Vacuum Manifold
Replace the parts and accessories on the Nalgene™ Vacuum Manifold with the Nalgene Vacuum Manifold Accessories.
Have Questions?
Catalog NumberStopper Number
DS0396-00808
Catalog number DS0396-0080
Price (USD)
44.55
Online exclusive
52.00
Save 7.45
Each
Stopper Number:
8
Price (USD)
44.55
Online exclusive
52.00
Save 7.45
Each
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
  • Compatible with Nalgene Vaccum Manifold (Cat. No. DS0345-0001)
  • Rubber stopper, number 8
  • 5/8 in. hole
Specifications
Diameter (Metric) Hole15.875 mm
For Use With (Equipment)Nalgene Vacuum Manifold (Cat. No. DS0345-0001)
MaterialRubber
Product TypeRubber Stopper
Stopper Number8
Unit SizeEach