Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter
Thermo Scientific™

Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter

Easily remove the Thermo ScientificTM NalgeneTM Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use.
Have Questions?
Catalog NumberCapacity (Metric)
DS3147-005035 mL
Catalog number DS3147-0050
Price (USD)
40.11
Online exclusive
80.68
Save 40.57
Case of 6
Capacity (Metric):
35 mL
Price (USD)
40.11
Online exclusive
80.68
Save 40.57
Case of 6
Easily remove the Thermo Scientific™ Nalgene™ Oak Ridge White Polycarbonate Conical Centrifuge Tube Adapter from rotors after use. This autoclavable adapter must be used with Nalgene 35L conical Oak Ridge Tubes (Cat. Nos. 3146, 3148).

Ordering Information:

To assure leakproof service, especially at speeds greater than 10,000 x g or when spinning hazardous materials, use Nalgene sealing cap assemblies DS3131-0020 or -0024.

Includes:

Supplied with linerless, polypropylene screw closure which will not leak during ordinary use.

Requires:

Both tubes require white polycarbonate tube adapter (see DS3147 0050 below) to centrifuge.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Specifications
No. per Case6
No. per Pack6
Product TypeCentrifuge Tube Adapter
ShapeConical
AutoclavableAutoclavable
Bottom ShapeConical
Capacity (Metric)35 mL
MaterialPolycarbonate
Max. RCF50,000 x g
Unit SizeCase of 6