Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Thermo Scientific™

Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature RangeArrayHolds
DS5116-00320°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
355501-20°C4 x 520 Tubes (0.2 to 0.5 mL)
5115-0012-20°C3 x 412 Tubes (0.2 to 0.5 mL)
5115-0032-20°C4 x 832 Tubes (0.2 to 0.5 mL)
5116-13000°C3 x 412 Tubes (12 to 13 mm dia.)
5116-16000°C3 x 412 Tubes (16 to 17 mm dia.)
DS5116-00120°C3 x 412 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
DS5114-0012-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
Catalog number DS5116-0032
Price (USD)
504.25
Online exclusive
661.78
Save 157.53
Each
Temperature Range:
0°C
Array:
4 x 8
Holds:
32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
Price (USD)
504.25
Online exclusive
661.78
Save 157.53
Each

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Array4 x 8
Description4x8 compartments, gel filled, 0 deg.
DesignGel-filled/white lid
Dimensions (L x W x H)9.56 x 6.19 x 5.75 in. (243 x 157 x 146 mm)
Height (English)5.75 in.
Height (Metric)146 mm
Holds Temp., Hours<1°C, 5.0
Length (English)9.56 in.
Length (Metric)243 mm
Width (English)6.19 in.
Width (Metric)157 mm
Capacity (Metric)0.2 to 0.5 mL
ColorBlue, Clear
Holds32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
MaterialPolycarbonate
Product TypeBenchtop Cooler
Temperature Range0°C
Unit SizeEach