Revco™ DxF -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes, 208-230V/60Hz, 12A - FAQs
Revco™ DxF -40°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes, 208-230V/60Hz, 12A - FAQs
View additional product information for Revco™ DxF -40°C Upright Ultra-Low Temperature Freezers - FAQs (DXF40040D)