How can I limit signal saturation when using the EnzChek Lysozyme Assay Kit (Cat. No. E22013)?
To limit signal saturation when using the EnzChek Lysozyme Assay Kit (Cat. No. E22013), try lowering the voltage (gain) on the PMT (photomultiplier tube). If you are still seeing signal saturation after lowering the voltage, try a shorter reaction time, and/or lower substrate concentration.