Escherichia coli (K-12 strain) BioParticles™, BODIPY™ FL conjugate - FAQs
Escherichia coli (K-12 strain) BioParticles™, BODIPY™ FL conjugate - FAQs
View additional product information for Escherichia coli (K-12 strain) BioParticles™, BODIPY™ FL conjugate - FAQs (E2864)
2 product FAQs found
What is the type of bond that attaches the dyes to the BioParticles probes?
We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.
What can the BioParticles product line be used for?
Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.