Escherichia coli BioParticles™ Opsonizing Reagent - FAQs
Escherichia coli BioParticles™ Opsonizing Reagent - FAQs
View additional product information for Escherichia coli BioParticles™ Opsonizing Reagent - FAQs (E2870)
2 product FAQs found
Are the Invitrogen BioParticles products sterile?
While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.
What is the type of bond that attaches the dyes to the BioParticles probes?
We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.