Selenite F Broth
Thermo Scientific™

Selenite F Broth

Selectively enrich Salmonella species from fecal, food, or environmental samples by inhibiting the growth of competing enteric bacteria.
Have Questions?
Catalog NumberFill Weight
EB1321E10.0 to 10.3 g
Catalog number EB1321E
Price (USD)
-
Fill Weight:
10.0 to 10.3 g
Selenite F Broth is used to selectively enrich Salmonella species from fecal, food, or environmental samples by inhibiting the growth of competing enteric bacteria.
Specifications
ColorStraw
Fill Weight10.0 to 10.3 g
FormatPlastic Universal Bottle
Packaging Type100 Plastic Universal Bottles with plastic screw cap closures in a box
Quantity100/Pk.
Shelf Life35 weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
SterilitySterile
Contents & Storage
2°C to 10°C