DH10B Competent Cells - FAQs
DH10B Competent Cells - FAQs
View additional product information for DH10B Competent Cells - FAQs (EC0113)
12 product FAQs found
Can I store my competent E. coli in liquid nitrogen?
We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.
How should I store my competent E. coli?
We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.
I'm getting overgrowth of colonies. Why?
Ensure that you are using the correct antibiotic at the appropriate concentration. Additionally, make sure the antibiotic is not expired. If colonies exhibit unexpected morphologies, contamination could be a factor. Check your S.O.C. medium and LB growth medium.
I'm only getting white colonies, but none of the clones have an insert. What can I do?
Here are a few suggestions:
- Small fragments/linkers are cloning in to your vector instead of your insert; to correct this, gel-purify the insert before ligation
- Ensure that the correct concentrations of X-gal and/or IPTG (if vector contains the lacIq marker) are used
- If spreading X-gal and/or IPTG on your plate, allow sufficient time for the reagents to diffuse into the plate
- Incubate your plate for a longer period to ensure full color development
I'm getting no colonies at all on my plates. Can you help?
We recommend trying the following:
- Carry out the puc19 transformation control; this gives you information about the performance of the cells.
- Check plates for expiration and correct media used (LB/agar).
- Confirm that the correct antibiotic and concentration was used.
What is the storage media for the competent cells?
Preparation procedures and formulations for all of our competent cells are proprietary. All chemically competent cells are delivered in an aqueous solution that contains a mixture of salts, along with a freezing stabilizer such as glycerol or DMSO.
What suggestions can you make for blue/white screening?
1. Use pUC or pUC-based vectors that contain the portion of the lacZ gene that allows for ? complementation.
2. Select an E. coli strain that carries the lacZdeltaM15 marker.
3. Plate transformations on plates containing X-gal. Spread 50 µg of 2% X-gal or 100 microliters of 2% bluo-gal (both can be dissolved in DMF or 50:50 mixture of DMSO:water) on the surface of a 100 mm plate and let dry. Alternatively, add directly to the cooled medium (~50 degrees C) before pouring the plates at a final concentration of 50 µg/mL for X-gal and 300 µg/mL for bluo-gal. Plates are stable for 4 weeks at 4 degrees C.
4. If the strain used carries the lacIq marker, add IPTG to induce the lac promoter. Spread 30 µl of 100 mM IPTG (in water) on 100 mm plates. Alternatively, add the IPTG directly to cooled medium (~50 degrees C) before pouring the plates to a final concentration of 1 mM. Plates are stable for 4 weeks at 4 degrees C.
5. Do not plate E. coli on medium containing glucose if using X-gal or bluo-gal for blue-white screening. Glucose competes as a substrate and prevents cells from turning blue.
Should I increase the heat shock time for my chemically competent cells during the transformation of a larger volume?
The recommended heat shock time does increase slightly with increasing volume of competent cells. For a 50 µl reaction volume, you should heat shock at 42°C for 30 seconds. For 100 µl, 45 seconds is recommended and for 250 µl, 60 seconds. It is important to do a positive control transformation of pUC19 along with transformation of your ligation product to accurately determine your relative efficiency of transformation.
Could the efficiency of my transformation in E. coli improve if I use more than the recommended amount of DNA?
It may be surprising, but in most cases transformation efficiency per µg of DNA will actually decrease when higher amounts of plasmid are transformed in one reaction. While you may see more colonies on your plates, much of the extra plasmid DNA you added will actually be wasted. Competent cells eventually become oversaturated with DNA, and adding more plasmid beyond that level will not result in any additional colonies. For example, when transforming 10 pg plasmid DNA, the efficiency of TOP10 cells is 1.0x10E9 colonies per µg of DNA that you added. If you transform 1 ng all at once, the overall efficiency is likely to decrease to ~1.0x 10E8 colonies per µg, and transforming 1 µg in a single reaction will likely result in efficiency less than 1.0 x 10E6 colonies per µg.
To maximize colony yield, it is better to transform smaller amounts of DNA in multiple reactions rather than adding all of the DNA to one reaction. This is most important when transforming a library, where you ideally want each plasmid to be represented by a colony after transformation.
I need a competent cell line that will generate as much DNA as possible from my vector of interest. What do you recommend?
The three most important things in this case are: endA- genotype, a high copy number origin of replication, and the culture scale. In addition, make sure (a) the culture is grown at 37 degrees C (if lower, the copy number can be lower), (b) try a super-rich media, like Terrific broth, (c) aerate the culture well (the volume of media should be no more than 10% of the volume of the flask), and (d) shake at 200-250 rpm.
What is the formulation of the SOC medium that is provided with competent cells?
SOC (Super Optimal Catabolite) Medium Preparation (for 1 Liter):
1) To a 2 Liter flask with stir bar add the following:
- Bacto Tryptone 20 g
- Yeast Extract 5 g
- Sodium Chloride (NaCl) 0.58 g
- Potassium Chloride (KCl) 0.186 g
2) Add sterile water to a final volume of 1 Liter.
3) Mix well on magnetic stir plate for 5-10 minutes or until all of the ingredients are well mixed and completely dissolved.
4) Autoclave 30 minutes.
5) Allow to cool to room temperature.
6) Add 10 ml of sterile 2M Magnesium Solution (1M Magnesium sulfate, 1M Magnesium chloride)and mix well.
7) Add 10 ml of sterile 2M Glucose and mix well. (Final Glucose concentration is 20 mM).
How can I obtain the highest possible transformation efficiency with Thermo Scientific DH5α and DH10B competent cells?
To achieve the highest possible transformation efficiency with Thermo Scientific DH5α (Cat. No. EC0112) and DH10B (Cat. No. EC0113) competent cells, please follow the transformation protocol provided with the product and pay attention to the following details:
- Make sure to use wet well-pressed ice to maximize cold surface volume in all the transformation steps.
- Perform the steps quickly and accurately.
- Use chilled microcentrifuge tubes and bury the whole tube up to the cap in the wet ice.
- Do not exceed the recommended heat shock time and transfer the tube with cells back to the wet ice immediately.