Novex™ TBE Gels, 6%
Invitrogen™

Novex™ TBE Gels, 6%

Novex™ TBE Gels 6% are designed to run on the XCell SureLock™ Mini-Cell and provide high-resolution analysis of restriction digests and PCR products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
EC6265BOX10-well
EC6261BOX1-well
EC62652BOX12-well
EC62655BOX15-well
Catalog number EC6265BOX
Price (USD)
237.65
Online exclusive
248.00
Save 10.35
Each
Wells:
10-well
Request bulk or custom format
Price (USD)
237.65
Online exclusive
248.00
Save 10.35
Each
Novex™ TBE Gels 6% provide high-resolution analysis of restriction digests and PCR products. Designed to run on the XCell SureLock™ Mini-Cell, the polyacrylamide gels give sharp, clearly resolved, intense bands, and provide separations of double-strand DNA fragments from 65–250 bp.

Novex TBE Gels are available in a variety of well formats and gel percentages and can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis.

Advantages of TBE Gels for nucleic acid separation:

  • High resolution and sensitivity with lower background staining
  • Requires ∼10% sample concentration and volume of large or agarose gels
  • Efficient blotting
  • Easy extraction of DNA from gels
  • Gel extraction does not interfere with enzymatic reactions
  • Accurate and reproducible results

Formulation

Novex TBE gels are manufactured with high-purity Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended buffers

For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionNovex TBE Gels, 6%, 10-well
For Use With (Equipment)XCell SureLock Mini-Cell
Quantity10 Gels/Box
Sample TypeDouble-stranded DNA (dsDNA)
Shipping ConditionWet Ice
Gel Percentage6%
Gel TypeTBE
Product TypeTBE Gel
Separation Range65 to 250 bp
Wells10-well
Unit SizeEach
Contents & Storage
• Catalog number refers to a single box of 10 gels

Store at 4°C.

Frequently asked questions (FAQs)

Can I stain my TBE gel? How?

Yes, you can stain your TBE gels with ethidium bromide, SYBR Green I, SYBR Green II, and the SilverXpress Silver Staining Kit. For ethidium bromide staining, soak the gel in a 2 µg/mL solution of ethidium bromide in ultrapure water for 20 minutes. Destain by rinsing with three successive 10-minute rinses of ultrapure water. Visualize bands under UV light.

What is the difference in composition between the Invitrogen 6% TBE Gels and the Invitrogen 6% DNA Retardation Gels?

The Invitrogen 6% DNA Retardation Gels contain 0.5X TBE. Both gels will work for gel retardation; however, the 1X TBE in the Invitrogen 6% TBE Gels have a higher ionic environment, which may affect DNA-protein interactions. The 0.5X TBE used in the Invitrogen 6% DNA Retardation Gels usually works better, as it offers good fragment separation in electrophoresis yet has an ionic strength low enough to promote DNA-protein interactions.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

What are the smallest fragments that can be visualized on the TBE gels?

On the Invitrogen 10% TBE gels, a 51 bp marker can be clearly seen. On the Invitrogen 20% TBE gel, the 18 and 12 bp markers can be clearly seen.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

What is the concentration of EDTA in the TBE gels? How about glycerol?

The EDTA concentration in our TBE gels is 0.06% (w/v). The 20% TBE gels contain 4% glycerol for maximal resolution. All other TBE gels contain 0.8% glycerol in a layer that represents the bottom 9% of the gel. There is no glycerol in the rest of the gel.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

Can I stain my TBE gel or my TBE-urea gel? How?

Yes, for ethidium bromide staining, soak the gel in a 2 µg/mL solution of ethidium bromide in ultrapure water for 20 minutes. Destain by rinsing with three successive 10-minute rinses of ultrapure water. Visualize bands under UV light

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

Citations & References (15)

Citations & References
Abstract
Prevalence and correlates of GB virus C infection in HIV-infected and HIV-uninfected pregnant women in Bangkok, Thailand.
Authors:Bhanich Supapol W, Remis RS, Raboud J, Millson M, Tappero J, Kaul R, Kulkarni P, McConnell MS, Mock PA, McNicholl JM, Roongpisuthipong A, Chotpitayasunondh T, Shaffer N, Butera S
Journal:J Med Virol
PubMed ID:21108337
'GB virus C (GBV-C) is an apathogenic virus that has been shown to inhibit HIV replication. This study examined the prevalence and correlates of GBV-C infection and clearance in three cohorts of pregnant women in Thailand. The study population consisted of 1,719 (1,387 HIV-infected and 332 HIV-uninfected) women from three ... More
An in vitro DNA double-strand break repair assay based on end-joining of defined duplex oligonucleotides.
Authors:Datta K, Purkayastha S, Neumann RD, Winters TA
Journal:Methods Mol Biol
PubMed ID:22941624
'DNA double-strand breaks (DSBs) are caused by endogenous cellular processes such as oxidative metabolism, or by exogenous events like exposure to ionizing radiation or other genotoxic agents. Repair of these DSBs is essential for the maintenance of cellular genomic integrity. In human cells, and cells of other higher eukaryotes, DSBs ... More
Smooth muscle phenotypic diversity is mediated through alterations in myocardin gene splicing.
Authors:Ilagan RM, Genheimer CW, Quinlan SF, Guthrie KI, Sangha N, Ramachandrannair S, Kelley RW, Presnell SC, Basu J, Ludlow JW
Journal:J Cell Physiol
PubMed ID:21792927
'Myocardin (MYOCD) is a smooth and cardiac muscle-specific transcriptional coactivator that is required for the proper expression of contraction-related genes. Through its function to transactivate effector genes, MYOCD plays an essential role in mediating the switch between contractile and non-contractile phenotypes, particularly in smooth muscle cells (SMC). There are at ... More
Genome wide full-length transcript analysis using 5' and 3' paired-end-tag next generation sequencing (RNA-PET).
Authors:Ruan X, Ruan Y
Journal:Methods Mol Biol
PubMed ID:22113299
'RNA-PET is a paired end tag (PET) sequencing method for full-length mRNA transcripts analysis using the next generation sequencer platforms such as Illumina GA and SOLiD. Unlike RNA-Seq method that sequences randomly sheared shotgun RNA short fragments, RNA-PET captures and sequences the 5'' and 3'' end tags of full-length cDNA ... More
Rapid, low-input, low-bias construction of shotgun fragment libraries by high-density in vitro transposition.
Authors:Adey A, Morrison HG, Asan Xun X, Kitzman JO, Turner EH, Stackhouse B, MacKenzie AP, Caruccio NC, Zhang X, Shendure J
Journal:Genome Biol
PubMed ID:21143862
We characterize and extend a highly efficient method for constructing shotgun fragment libraries in which transposase catalyzes in vitro DNA fragmentation and adaptor incorporation simultaneously. We apply this method to sequencing a human genome and find that coverage biases are comparable to those of conventional protocols. We also extend its ... More