Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations
Actual product may vary
Thermo Scientific™
Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Stations with these crimper and decrimper sets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeType
ECMJ-88 mmElectronic Crimper
ECMJ-1111 mmElectronic Crimper
ECDJ-1111 mmElectronic Decrimper
ECMJ-1313 mmElectronic Crimper
ECDJ-1313 mmElectronic Decrimper
ECMJ-13FO13 mmElectronic Crimper (Flip-off)
ECMJ-2020 mmElectronic Crimper
ECDJ-2020 mmElectronic Decrimper
ECMJ-20FO20 mmElectronic Crimper (Flip-off)
Catalog number ECMJ-8
Price (USD)
489.49
Each
Size:
8 mm
Type:
Electronic Crimper
Price (USD)
489.49
Each
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Station with these crimper and decrimper sets.
Available in a variety of sizes and formats.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)Programmable Electronic High-Power Crimp Station, 8 mm Vial Closure
No. per Pack1 Ea.
Size8 mm
TypeElectronic Crimper
Unit SizeEach