Eutech™ EcoTestr pH2 pH meter
Eutech™ EcoTestr pH2 pH meter
Thermo Scientific™

Eutech™ EcoTestr pH2 pH meter

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
카탈로그 번호Measurement Modes
ECOPHTEST2pH
카탈로그 번호 ECOPHTEST2
제품 가격(KRW)
-
Measurement Modes:
pH

From the market leader of testers comes the EcoTestr pH 1 & pH 2 – a new range of value-for-money, quality pH tester series designed for fuss-free pH measurements. Accurate, and easy to use, these testers are ideal for quick pH checks in hydroponic gardening, aquaculture, agriculture, pools, simple lab work and other water/wasterwater applications.

Features:

  • 0.0 to 14.0 full pH range
  • Auto-calibration & auto-buffer recognition
  • IP67 waterproof – tester floats on water for easy retrieval
  • Long sensor life – no need to change sensors
  • Click-lock battery compartment requires no tools for battery replacements
  • Hold function freezes reading temporarily for ease of recording
  • Sensor protective cap that doubles up as a container for samples or calibration solution

Applications

Aquariums & aquaculture, hydroponics, swimming pools, spas & hot tubs, lab work and education, wastewater treatment, environment monitoring

Specifications

  • pH Range: 0.0 to 14.0 pH
  • Resolution: 0.1 pH
  • Accuracy: ±0.1 pH
  • pH Calibration Points: 3 (Auto)
  • Operating Temperature: 0 to 50°C
  • Non-Volatile Memory: Yes
  • Hold Function: Yes
  • Auto-Off: 8.5 mins after last key pressed
  • LCD Display: 3.5 digit with annunciator
  • Power: 4 X 1.5 V A76 Micro alkaline batteries
  • Ingress Rating: IP67 Waterproof
사양
정확도±0.1 pH
정확도(pH)±0.1
설명Waterproof pHTestr 2 Testr with ATC; 3 point Calibration
치수(길이 x 폭 x 높이)Boxed: 24.5 cm x 13.5 cm x 4.5 cm
메모리Yes, non-volatile
온도 보상ATC
작동 온도(미터법) 작동0°C to 50°C
중량(미터법)137 g
보정Automatic
Calibration Points3
Measurement ModespH
범위(pH)0 to 14
분해능(pH)0.1
Unit SizeEach