Alanine Aminotransferase (ALT/GPT) Activity Kit
Invitrogen™

Alanine Aminotransferase (ALT/GPT) Activity Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Catalog NumberQuantity
EEA00196 tests
Catalog number EEA001
Price (JPY)
116,700
Each
Quantity:
96 tests

The Alanine Aminotransferase (ALT/GPT) Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ALT activity in animal serum, plasma, tissue, cultured cells, and cell culture supernatant. ALT is an important pyridoxal phosphate dependent enzyme in the intermediate metabolism of glucose and proteins. Clinically, the activity of serum alanine aminotransferase is often used as a marker for alcoholic liver disease, liver cirrhosis, and acute viral hepatitis.

In this assay, ALT catalyzes the amino conversion reaction between alanine and α-ketoglutaric acid to produce pyruvic acid and glutamic acid. Phenylhydrazine is added to form phenylhydrazone with pyruvic acid. Phenylhydrazone is a reddish brown solution under alkaline conditions, and ALT activity can be calculated by measuring the OD values at 510 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.75–72.3 IU/L
Assay Sensitivity0.75 IU/L
For Use With (Equipment)Microplate Reader
Incubation Time50 min.
Interassay CV9
Intraassay CV5.3
Quantity96 tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedAlanine Aminotransferase (ALT/GPT)
Total Assay Time80 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.