Aspartate Aminotransferase (AST/GOT) Activity Kit
Invitrogen™

Aspartate Aminotransferase (AST/GOT) Activity Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA00396 tests
Catalog number EEA003
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Aspartate Aminotransferase (AST/GOT) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of aspartate aminotransferase activity in various sample types. AST/GOT is a key enzyme in nitrogen metabolism, which is widely found in plasma and body tissues, including liver, heart, skeletal muscle, kidney, brain, pancreas, lung, and erythrocytes. Changes in AST/GOT activity were found in acute pancreatitis, ischemic stroke, severe burns, periodontitis, acute renal disease, and motor neuron disease.

The assay is based on the ability of AST/GOT to enable alpha-ketoglutaric acid and aspartic acid to displace amino and keto groups to form glutamic acid and oxaloacetic acid. Oxaloacetic acid can decarboxylate itself to form pyroracemic acid during the reaction. Pyroracemic acid reacts with 2,4-dinitro phenyl hydrazine (DNPH) to form 2,4, dinitrophenylhydrazone, showing reddish-brown color in alkaline solution. The OD values can be measured to calculate the enzyme activity.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range1.1–72.3 IU/L
Assay Sensitivity1.1 IU/L
For Use With (Equipment)Microplate Reader
Incubation Time50 min.
Interassay CV6.8
Intraassay CV5.3
Quantity96 tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedAspartate Aminotransferase (AST/GOT)
Total Assay Time80 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.