Lactate Dehydrogenase (LDH) Colorimetric Activity Kit
Invitrogen™

Lactate Dehydrogenase (LDH) Colorimetric Activity Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA01396 tests
Catalog number EEA013
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Lactate Dehydrogenase (LDH) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of LDH in serum, plasma, tissue, and cell culture samples. LDH is an enzyme that catalyzes the conversion of lactate to pyruvate, an essential step in cellular energy metabolism. Elevated LDH levels are associated with tissue damage, inflammation, and certain diseases, making it a useful biomarker in clinical and research settings.

In this assay, LDH catalyzes lactic acid to produce pyruvate. Pyruvate reacts with 2, 4-dinitrophenylhydrazine to form pyruvate dinitrophenylhydrazone, which is red-brown in alkaline solution. The color intensity is proportional to pyruvate concentration, where the activity of LDH can be calculated by measuring OD value.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range6–1000 U/L
Assay Sensitivity6 U/L
For Use With (Equipment)Microplate Reader
Incubation Time35 min.
Interassay CV2.4
Intraassay CV1.8
Quantity96 tests
Sample Volume40 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedLactate Dehydrogenase (LDH)
Total Assay Time45 min.
Detection MethodColorimetric
Sample TypeHydrothorax, Plasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.