Low-density Lipoprotein Cholesterol (LDL-C) Colorimetric Assay Kit
Invitrogen™

Low-density Lipoprotein Cholesterol (LDL-C) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA01496 tests
Catalog number EEA014
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Low-Density Lipoprotein Cholesterol (LDL-C) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of LDL-C in serum, plasma, tissue, cells, and cell culture supernatant samples. LDL-C is the main lipoprotein in fasting plasma, accounting for about two-thirds of plasma lipoproteins, and is the main vehicle for transporting cholesterol to extrahepatic tissues. The content of LDL-C is related to the incidence of cardiovascular disease and the degree of lesions and is considered to be the main pathogenic factor of atherosclerosis.

The assay is based on the reaction of LDL-C with enzyme reagents containing cholesterol esterase and cholesterol oxidase to produce hydrogen peroxide. Hydrogen peroxide is catalyzed by an oxidase in the presence of 4-aminoantipyrine (4-AA) and phenol (T-OOS) to form a red quinone compound with a maximum absorption peak at 546 nm. The LDL-C content in the sample can be calculated by measuring the OD value.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.2–12 mmol/L
Assay Sensitivity0.2 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time10 min.
Interassay CV10
Intraassay CV5.5
Quantity96 tests
Sample Volume2.5 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedLow-density Lipoprotein Cholesterol (LDL-C)
Total Assay Time20 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.