Malondialdehyde (MDA) Colorimetric Assay Kit
Invitrogen™
Malondialdehyde (MDA) Colorimetric Assay Kit
These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Número de catálogoCantidad
EEA01596 pruebas
Número de catálogo EEA015
Precio (MXN)
15,765.19
Each
Cantidad:
96 pruebas
Pedido a granel o personalizado
Precio (MXN)
15,765.19
Each
The Malondialdehyde (MDA) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of MDA in serum, plasma, tissue, and other samples. MDA is a biomarker of oxidative stress and lipid peroxidation, and detection of MDA in samples can indirectly reflect the level of cellular damage from oxygen free radicals.
In this assay MDA, as the catabolite of lipid peroxide, reacts with thiobarbituric acid (TBA) to produce a red compound. This compound has a maximum absorption peak at 532 nm, which can be used to measure the amount of malondialdehyde present.
Key features
  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Especificaciones
Intervalo del ensayo2.92–40 μmol/L
Sensibilidad del ensayo1.13 μmol/L
Para utilizar con (equipo)Lector de microplacas
Tiempo de incubación40 min.
CV interensayo7.2
CV intraensayo4.1
Cantidad96 pruebas
Volumen de muestra40 μL
Condiciones de envíoDry Ice, 2–8°C
Nombre del kit de destinoMalondialdehyde (MDA)
Tiempo total del ensayo65 min.
Método de detecciónColorimétrico
Tipo de muestraPlasma, Serum, Tissue Homogenates
Unit SizeEach
Contenido y almacenamiento
Store at 2–8°C.