Myeloperoxidase (MPO) Colorimetric Activity Kit
Invitrogen™

Myeloperoxidase (MPO) Colorimetric Activity Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA01696 tests
Catalog number EEA016
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Myeloperoxidase (MPO) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of MPO in serum, plasma, milk, and animal tissue. MPO is an enzyme that belongs to the heme peroxidase family and is found in various immune cells, particularly neutrophils. It plays a crucial role in the innate immune response and is involved in host defense mechanisms against pathogens. MPO is closely related to the pathogenesis of diseases such as cardiovascular disease, lung injury, and cancer.

In this assay MPO reduces hydrogen peroxide to a complex that reacts with o-dianisidine to produce a yellow product that has a maximum absorption peak at 460 nm, which can be used to measure the amount of MPO present.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range19.42–893.31 U/L
Assay Sensitivity19.42 U/L
For Use With (Equipment)Microplate Reader
Incubation Time40 min.
Interassay CV7.3
Intraassay CV5.4
Quantity96 tests
Sample Volume40 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedMyeloperoxidase (MPO)
Total Assay Time65 min.
Detection MethodColorimetric
Sample TypePlasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.