Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit
Invitrogen™

Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02196 tests
Catalog number EEA021
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Thiobarbituric Acid Reactants (TBARS) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of TBARS in serum, plasma, animal tissue, cultured cells, and other sample types. TBARS are products of lipid peroxidation, a process that occurs when polyunsaturated fatty acids in cellular membranes are attacked by oxygen free radicals. TBARS, particularly malondialdehyde (MDA), are commonly used as biomarkers of lipid peroxidation and can reflect the degree of oxidative stress and cellular injury in organisms.

In this assay, TBARS and thiobarbituric acid (TBA) react under high temperature and acidic conditions. This reaction forms a pink compound, which has an optical density measured at 530–540 nm and the concentration of which is linearly related to the concentration of TBARS in the sample.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range2.6–100 μmol/L
Assay Sensitivity0.85 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time60 min.
Interassay CV7.1
Intraassay CV4.3
Quantity96 tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedThiobarbituric Acid Reactants (TBARS)
Total Assay Time80 min.
Detection MethodColorimetric
Sample TypePlasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.