Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit
Invitrogen™

Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02296 tests
Catalog number EEA022
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, whole blood, tissue, cells, and culture supernatant. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.

The assay quantifies the ability of the sample to reduce ferric ions (Fe3+) to ferrous ions (Fe2+) using a phenanthroline substance. The resulting complex exhibits a characteristic absorbance at 520 nm, which is directly proportional to the T-AOC of the sample.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.62–190.43 U/mL
Assay Sensitivity0.62 U/mL
For Use With (Equipment)Microplate Reader
Incubation Time40 min.
Interassay CV5.6
Intraassay CV4.8
Quantity96 tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedTotal Antioxidant Capacity (T-AOC)
Total Assay Time50 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Blood, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.