Triglyceride (TG) Colorimetric Assay Kit
Invitrogen™

Triglyceride (TG) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02896 tests
Catalog number EEA028
Price (USD)
710.00
Each
Quantity:
96 tests
Request bulk or custom format
Price (USD)
710.00
Each
The Triglyceride (TG) Colormetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of triglycerides in serum, plasma, and tissue samples. Triglycerides are a type of lipid molecule that play a crucial role in energy storage and metabolism. They are the main component of vegetable oil, animal fat, low-density lipoprotein (LDL), and very low-density lipoprotein (VLDL). Elevated levels of triglycerides in the blood have been associated with an increased risk of cardiovascular disease, glucose metabolism, insulin resistance, and Type 2 diabetes.

The assay utilizes the GPO-PAP method, which involves the hydrolysis of triglycerides to glycerol and fatty acids. Glycerol is then oxidized by glycerol kinase to glycerol-3-phosphate, which is oxidized by GPO and produces hydrogen peroxide. In the presence of 4-aminoantipyrine and phenol, hydrogen peroxide is catalyzed by peroxidase (POD) to produce colored quinones which can be measured at 510 nm to calculate the amount of TG in the sample.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits—across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.14–10 mmol/L
Assay Sensitivity0.14 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time10 min.
Interassay CV9.2
Intraassay CV4.1
Quantity96 tests
Sample Volume2.5 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedTriglyceride (TG)
Total Assay Time30 min.
Detection MethodColorimetric
Sample TypeSerum, Plasma, Tissue
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.