L-Lactic Acid (LA) Colorimetric Assay Kit
Invitrogen™

L-Lactic Acid (LA) Colorimetric Assay Kit

With this kit, LDH catalyzes the conversion of both lactate and NAD+ into pyruvic acid and NADH respectively.
Have Questions?
Catalog NumberQuantity
EEA037100 Tests
Catalog number EEA037
Price (USD)
696.00
Each
Quantity:
100 Tests
Price (USD)
696.00
Each

With this kit, LDH catalyzes the conversion of both lactate and NAD+ into pyruvic acid and NADH respectively. 1-Methoxy-5-methyl phenazine methyl sulfate (PMS) transfers hydrogen from NADH to NBT, which deoxidizes into a purple chromogenic substrate. Lactic acid content can be calculated by measuring the OD value at 530 nm. This kit is useful for researchers studying lactic acid metabolism and cellular respiration.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.05 to 6.0 mmol/L
Assay Sensitivity0.05 mmol/L
For Use With (Equipment)Spectrophotometer
Incubation Time10 min.
Interassay CV1.9%
Intraassay CV1.1%
Product TypeKit
Quantity100 Tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedL-Lactic Acid (LA)
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Tissue, Cells, Cell Culture Supernatant
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.