Ascorbate Peroxidase (APX) Activity Assay Kit
Invitrogen™

Ascorbate Peroxidase (APX) Activity Assay Kit

With this kit, ascorbate peroxidase (APX) catalyzes a reaction between ascorbic acid (ASA) and hydrogen peroxide (H2O2), with ASA being oxidized to monodehydroascorbic acid (MDASA).
Have Questions?
Catalog NumberQuantity
EEA04096 Tests
Catalog number EEA040
Price (USD)
696.00
Each
Quantity:
96 Tests
Price (USD)
696.00
Each

With this kit, ascorbate peroxidase (APX) catalyzes a reaction between ascorbic acid (ASA) and hydrogen peroxide (H2O2), with ASA being oxidized to monodehydroascorbic acid (MDASA). The absorbance of the solution at 290 nm declines as the oxidation of ASA progresses. APX activity can be calculated by measuring the decrease in absorbance at 290 nm. This kit is useful for researchers studying antioxidant enzymes and oxidative stress.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.071 to 47 U/g tissue
Assay Sensitivity0.071 U/g tissue
For Use With (Equipment)spectrophotometer
Incubation Time1 min.
Interassay CV6.4%
Intraassay CV4.8%
Product TypeKit
Quantity96 Tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedAscorbate Peroxidase (APX)
Total Assay Time60 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypePlant Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.