NAD<sup>+</sup>/NADH Colorimetric Assay Kit
Invitrogen™

NAD+/NADH Colorimetric Assay Kit

NAD+ and NADH are coenzymes that transfer electrons during REDOX reactions and can be used as cofactors of many enzymes to participate in intracellular reactions.
Have Questions?
Catalog NumberQuantity
EEA04296 Tests
Catalog number EEA042
Price (USD)
696.00
Each
Quantity:
96 Tests
Price (USD)
696.00
Each

Detection of total content of NAD+ and NADH

With this kit, ethanol generates acetaldehyde under the action of an enzyme. Meanwhile, NAD+ is reduced to NADH. NADH, under the action of hydrogen transmitter, transfers electrons to WST-8 to produce a yellow product that has a characteristic absorption peak at 450 nm. The total content of NAD+ and NADH can be quantified by measuring the OD value at 450 nm.

Detect NADH

After treating the sample, it is heated at 60°C. The NAD+ of the sample is decomposed and only NADH remains. NADH reduces WST-8 to form a yellow product, and the amount of NADH is determined by measuring the OD value at 450 nm.

Detect NAD+ and NAD+/NADH

The content of NAD+ and the ratio of NAD+/NADH in the sample can be obtained according to the total content of NAD+ and NADH obtained from the first two steps as well as the separate content of NADPH.

Note: NADP+ and NADPH have no effect on the determination results.

This kit is useful for researchers studying cellular metabolism and redox biology.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.02 to 5.0 μmol/L
Assay Sensitivity0.02 μmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time30 min.
Interassay CV9.1%
Intraassay CV1.8%
Product TypeKit
Quantity96 Tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedNAD+/NADH
Total Assay Time70 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeAnimal Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.