Hydroxyproline (HYP) Colorimetric Assay Kit
Invitrogen™

Hydroxyproline (HYP) Colorimetric Assay Kit

With this kit, the sample is hydrolyzed to generate free hydroxyproline (HYP), and hydroxyproline produces an oxidation product under the action of an oxidizing agent.
Have Questions?
Catalog NumberQuantity
EEA047100 Tests
Catalog number EEA047
Price (USD)
696.00
Each
Quantity:
100 Tests
Price (USD)
696.00
Each

With this kit, the sample is hydrolyzed to generate free hydroxyproline (HYP), and hydroxyproline produces an oxidation product under the action of an oxidizing agent. The generated oxidation product reacts with a chromogenic agent to produce a burgundy color. The concentration of hydroxyproline can be calculated by measuring the OD value at 558 nm. This kit is useful for researchers studying collagen metabolism and tissue remodeling.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.032 to 10 μg/mL
Assay Sensitivity0.032 μg/mL
For Use With (Equipment)spectrophotometer
Incubation Time30 min.
Interassay CV5.7%
Intraassay CV4.9%
Product TypeKit
Quantity100 Tests
Sample Volume400 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedHydroxyproline (HYP)
Total Assay Time45 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Urine, Animal Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.