Citrate Synthase(CS) Activity Assay Kit
Invitrogen™

Citrate Synthase(CS) Activity Assay Kit

With this kit, citrate synthetase (CS) catalyzes acetyl-CoA and oxaloacetate to produce limonyl-CoA, which can further produce CoA.
Have Questions?
Catalog NumberQuantity
EEA05496 Tests
Catalog number EEA054
Price (USD)
696.00
Each
Quantity:
96 Tests
Price (USD)
696.00
Each

With this kit, citrate synthetase (CS) catalyzes acetyl-CoA and oxaloacetate to produce limonyl-CoA, which can further produce CoA. The yellow product generated by this reaction has a characteristic absorption peak at 412 nm. The activity of CS can be calculated by measuring the change of absorbance value at 412 nm. This kit is useful for researchers studying the citric acid cycle and energy metabolism.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range2.09 to 110 U/L
Assay Sensitivity2.09 U/L
For Use With (Equipment)Microplate Reader
Incubation Time8 min.
Interassay CV7.9%
Intraassay CV5.0%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedCitrate Synthase(CS)
Total Assay Time15 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeAnimal Tissue, Plant Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.