ATP Colorimetric Assay Kit
Invitrogen™

ATP Colorimetric Assay Kit

Adenosine triphosphate (ATP) is a high-energy phosphate compound that is the most direct energy source in the body, and its content is directly related to the state of energy metabolism in the body. With this kit, hexokinase (HK) catalyzes the generation of glucose and ATP into glucose-6-phosphate.
Have Questions?
Catalog NumberQuantity
EEA060100 Tests
Catalog number EEA060
Price (USD)
696.00
Each
Quantity:
100 Tests
Price (USD)
696.00
Each

Adenosine triphosphate (ATP) is a high-energy phosphate compound that is the most direct energy source in the body, and its content is directly related to the state of energy metabolism in the body. With this kit, hexokinase (HK) catalyzes the generation of glucose and ATP into glucose-6-phosphate. Glucose-6-phosphate dehydrogenase (G6PD) further catalyzes the dehydrogenation of glucose-6-phosphate into NADPH. NADPH then reacts with a color developer, and the product has a characteristic absorption peak at 460 nm that is used to determine the ATP content. This kit is useful for researchers studying cellular energy metabolism and bioenergetics.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.72 to 40.00 μmol/L
Assay Sensitivity0.72 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time5 min.
Interassay CV3.6%
Intraassay CV1.9%
Product TypeKit
Quantity100 Tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedATP
Total Assay Time10 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeAnimal Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.