Human FGF4 (Cell Lysates) ELISA Kit - FAQs
Human FGF4 (Cell Lysates) ELISA Kit - FAQs
View additional product information for Human FGF4 (Cell Lysates) ELISA Kit - FAQs (EHFGF4CL, EHFGF4CLX10, EHFGF4CLX5)