IL-17A Human ELISA Kit
Invitrogen™

IL-17A Human ELISA Kit

The Human IL17A ELISA research-use-only kit is to be used for the quantitative determination of IL17A in samples (see sampleRead more
Have Questions?
Catalog NumberQuantity
EHIL17A1 Plate Kit
Catalog number EHIL17A
Price (EUR)
-
Quantity:
1 Plate Kit
The Human IL17A ELISA research-use-only kit is to be used for the quantitative determination of IL17A in samples (see sample types and species reactivity indicated) using 96-well plates and a microplate reader. The assay will recognize both natural and recombinant forms of this target.

Performance characteristics

• Sensitivity: 5 pg/mL
• Standard curve range: 31.25 pg/mL - 2000 pg/mL
• Sample types: serum, plasma, cell culture supernatant
• Specificity: This ELISA is specific for the measurement of natural and recombinant human IL17A. No significant crossreaction with human IL17B, IL17D, IL17E, IL17F, IL2, IL4, IL5, IL6, IL8, IL10, IL12, IL13, IL16 or mouse IL17A, IL1A, IL1B or GMCSF was found.
• Sample volume: 50 μL
• Total assay incubation time: 3.5 hours

Principle of the method
The Thermo Scientific™ Pierce™ Human IL17A ELISA is a solid-phase, sandwich-type enzyme-linked immunosorbent assay (ELISA) designed to measure the amount of the target between a 'sandwich' or two layers of antibodies. A target-specific antibody has been pre-coated onto the wells of the microtiter plate provided. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Background information
IL17A, also known as Interleukin 17, is a proinflammatory cytokine secreted by activated memory CD4+ T-cells. IL17A is noted for involvement in chronic inflammatory diseases, including psoriasis, multiple sclerosis and rheumatoid arthritis.

Related links:
Learn more about ELISA kits.
Learn more about other immunoassays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeSee product documentation
Assay SensitivitySee product documentation
DescriptionHuman IL-17A ELISA Kit
For Use With (Equipment)Microplate Reader
Former IdentifiersPierce
Geneinterleukin 17A
Gene AliasCtla-8, Ctla8, IL-17, IL-17A, Il17
Gene ID (Entrez)3605
Gene SymbolIL17A
Label or DyeHRP
ProteinInterleukin-17A
Protein AliasesIL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8
Protein FamilyCytokines & Receptors
Quantity1 Plate Kit
Target Kit NamedIL-17A
Target Species ValidatedHuman
UniProt IDQ16552
Detection MethodColorimetric
Immunoassay Kit FormatCoated ELISA Kit
Target SpeciesHuman
Unit Size96 tests
Contents & Storage
Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Frequently asked questions (FAQs)

Will ProcartaPlex multiplex assays give me the same results for each analyte as my current ELISA tests?

ProcartaPlex multiplex assays, which are based on Luminex xMAP technology, provide a versatile platform that gives users more flexibility and a greater array of options for analyte detection. Whether you are testing for single or multiple analytes, ProcartaPlex multiplex assays deliver accurate analytical performance using efficient, easy-to-follow protocols. Each of these assays has undergone the same development, validation, manufacturing, and quality control standardization we conduct for our ELISAs. Each lot of ProcartaPlex multiplex assays as well as ELISA assays is fully qualified with the appropriate sample type (i.e., species-specific serum, plasma, and cell culture supernatants), and each lot is evaluated based on the following performance characteristics:

Specificity-each analyte is screened to make sure there is no significant cross-reactivity with other analytes in the multiplex test
Sensitivity-each analyte is evaluated for both functional sensitivity (differentiation from background) and lower limit of detection (LLOD)
Precision/accuracy-multiplex assays have good intra-assay precision (<10% CV), inter-assay precision (<10% CV), and lot-to-lot consistency (<20% CV); these values are comparable to or better than most ELISA tests
ProcartaPlex multiplex assays are regularly tested against the matching ELISAs. Therefore, you can switch easily from ProcartaPlex assays to ELISA and vice versa with reliable results. Most of our ProcartaPlex assays use the same antibody pairs as our traditional plate-based ELISAs, resulting in high correlation (R2 > 0.9) between the two assays.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Why should I consider switching from ELISA technology to multiplexing?

ELISA is a simple and powerful way to quantify individual proteins specifically in complex samples. The selectivity of ELISA is achieved through the use of qualified single- or double-antibody sandwich technology, and accurate quantitation is achieved through the use of calibrated standards. ELISAs can detect low-level proteins and can be performed in a 96-well format with only 60 minutes of hands-on time. In addition, the results obtained with ELISAs are generally very reproducible. While ELISA has been established as a standard method of protein analysis, multiplexing methods that enable the measurement of multiple analytes simultaneously in a single sample address a number of specific limitations:

ELISA allows for the measurement of only one analyte at a time in a given sample, limiting investigators' increasing need to measure multiple targets in their research studies.
The low available volume of many samples being studied may limit the number of times analyses can be conducted. This is especially true in small animal research, in pediatric testing, and in microplate assays providing limited sample volumes. The ability to assay multiple analytes in a single small-volume sample enables more effective use of each sample.
Difficulties in data interpretation can arise when comparing analyte levels measured by multiple ELISAs, each assay having been performed with different sample aliquots and each susceptible to systematic errors leading to decreased precision and accuracy.
Many analytes require assays with broad dynamic ranges to avoid repeat testing or out-of-range values. Multiplex assays can be designed to have large dynamic ranges for all of the analytes, or ranges tailored to various expected analyte concentrations.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

I performed an ELISA assay, and the A450 readings for my duplicate wells were very inconsistent. What could have gone wrong?

Here are possible causes and solutions:

Errors in pipetting the standards or samples or in subsequent steps. Always dispense into wells quickly and in the same order. Do not touch the pipette tip on the individual microwells when dispensing. Use calibrated pipettes and the appropriate tips for that device. Check for any leaks in the pipette tip.
Repetitive use of tips for several samples or different reagents. Use fresh tips for each sample or reagent transfer.
Wells have been scratched with the pipette tip or washing tips. Use caution when dispensing into and aspirating out of microwells.
Liquid transferred from well to well during incubations. Adjust the orbital shaker or check for correct rotator rpm. Peel the adhesive plate cover off carefully.
Incorrect volumes of materials dispensed into the microwells. Follow the protocol for dispensing volumes of reagents. Check calibration of the pipettes.
Standard diluted with the serum, culture medium, or other buffer. Dilute the standard with the standard diluent buffer provided in the kit.
Particulates or precipitates present in the samples. Remove any particulates/precipitates by centrifugation prior to dispensing into the assay.
Dirty microwells: visible debris within or on bottom of microwells. Inspect the microwells and invert the plate to remove debris. Wipe the bottom of the plate with an absorbent tissue after each wash step. Never insert tissue into the microwells.
“Edge effect” due to uneven temperature between the outer-edge wells and the wells in the center of the plate. Seal the plate completely with a cover during incubations, and place the plate in the center of the incubator when 37 degrees C incubation is indicated.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

I got a poor standard curve after my ELISA. Why is this?

Here are possible causes and solutions:

Improper preparation of standard stock solution.Dilute the lyophilized standard as directed on the vial label, only with the standard diluent buffer or a diluent that most closely matches the matrix of your sample.
Reagents (lyophilized standard, standard diluent buffer, etc.) from different kits, with either different analytes or different lot numbers, were substituted. Never substitute any components from another kit.
Errors in pipetting the standard or in subsequent steps. Always dispense into wells quickly and in the same order. Do not touch the pipette tips on the individual microwells when dispensing. Use calibrated pipettes and the appropriate tips for that device.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

I see very weak to no color development after my ELISA. What happened?

Here are possible causes and solutions:

Reagents not at room temperature (approximately 25 plus or minus 2 degrees C) at start of assay. Allow all reagents to warm to room temperature prior to commencing the assay.
Incorrect storage of components, e.g., not stored at 2-8 degrees C. Store all components exactly as directed in the protocol and on labels.
Anti-rabbit IgG HRP or streptavidin-HRP working solution made more than 15 minutes before use in assay. Use the diluted anti-rabbit IgG HRP or streptavidin-HRP within 15 minutes of dilution.
Expired reagents.Check expiration dates upon receipt of kit and use the kit prior to expiration.
Plate read at incorrect wavelength. The correct wavelength to read ELISAs using the TMB substrate is 450 nm.
TMB solution lost activity. Ensure that the TMB solution is clear before it is dispensed into the plate wells. A blue color and/or the presence of particulate matter indicate that the product is contaminated. Please contact Technical Support if this problem is noted. To avoid contamination, we recommend that the quantity required for an assay be dispensed into a previously unused disposable trough for pipetting. Discard any TMB solution left in the trough and do not put it back in the bottle. Avoid contact between the TMB solution and items containing metal ions. Do not cover your plates with aluminum foil or aluminum-coated Mylar sheets because this can cause color development in the absence of HRP.
Attempt to measure analyte in a matrix for which the ELISA assay is not optimized. Contact Technical Support when using alternative sample types.
Wells have been scratched with pipette tip or washing tips. Use caution when dispensing into and aspirating out of microwells.
Incorrect chromogen or stop solution used. Use only the chromogen and stop solution supplied with the kit.
Standard diluent buffer added to all wells rather than the designated wells. Follow the protocol and only add the standard diluent to the designated wells and to the samples where it is required, or to samples producing signals greater than that of the highest standard.
Use of buffer containing azide, which is not compatible with HRP. Avoid the use of azide in the assay.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.