Human PF4 ELISA Kit - FAQs
Human PF4 ELISA Kit - FAQs
View additional product information for Human PF4 ELISA Kit - FAQs (EHPF4X10, EHPF4, EHPF4X5)
1 product FAQs found
What is the plate format for PF4 Human ELISA Kit?
PF4 Human ELISA Kit includes a pre-coated 96 well plate, 12 x 8 strips.